Search
Search

LOC4348697| Gene Symbol: LOC4348697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759348.2 | XP_015614834.1 | 1 - 2 | 435 | 121 | |
| GenBank mRNA | AK072305.1 | - | 1 - 2 | 266 | 121 | |
| AK106265.1 | - | 1 - 2 | 303 | 121 | ||
Set Membership: |
|