Search
Search

LOC4349151| Gene Symbol: LOC4349151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759179.2 | XP_015614665.1 | 5 - 6 | 702 | 106 | |
| XM_015759180.2 | XP_015614666.1 | 5 - 6 | 702 | 106 | ||
| GenBank mRNA | AK072155.1 | - | 4 - 5 | 2089 | 106 | |
Set Membership: |
|