Search
Search

LOC4325831| Gene Symbol: LOC4325831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795620.2 | XP_015651106.1 | 1 - 2 | 170 | 77 | |
| GenBank mRNA | AK071109.1 | - | 1 - 2 | 118 | 77 | |
| AK104139.1 | - | 1 - 2 | 113 | 77 | ||
| AK104321.1 | - | 1 - 2 | 113 | 77 | ||
| AK119902.1 | - | 1 - 2 | 112 | 77 | ||
Set Membership: |
|