Search
Search

LOC4329811| Gene Symbol: LOC4329811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768036.2 | XP_015623522.1 | 2 - 3 | 392 | 59 | |
| GenBank mRNA | AK069595.1 | - | 2 - 3 | 392 | 59 | |
Set Membership: |
|