Search
Search

LOC4331772| Gene Symbol: LOC4331772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775325.2 | XP_015630811.1 | 3 - 4 | 1414 | 80 | |
| XM_015775326.2 | XP_015630812.1 | 3 - 4 | 1411 | 80 | ||
| XM_015775327.2 | XP_015630813.1 | 3 - 4 | 1031 | 80 | ||
| GenBank mRNA | AK242042.1 | - | 4 - 5 | 1070 | 80 | |
| AK243489.1 | - | 3 - 4 | 1353 | 80 | ||
Set Membership: |
|