Search
Search

LOC4344922| Gene Symbol: LOC4344922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792567.2 | XP_015648053.1 | 2 - 3 | 1034 | 78 | |
| GenBank mRNA | AK119306.1 | - | 3 - 4 | 903 | 78 | |
Set Membership: |
|