Search
Search

LOC4346998| Gene Symbol: LOC4346998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757132.2 | XP_015612618.1 | 3 - 4 | 1076 | 67 | |
| GenBank mRNA | JN608807.1 | - | 3 - 4 | 998 | 67 | |
Set Membership: |
|