Search
Search

LOC4351057| Gene Symbol: LOC4351057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761261.2 | XP_015616747.1 | 1 - 2 | 592 | 73 | |
| GenBank mRNA | AK107017.1 | - | 1 - 2 | 592 | 73 | |
Set Membership: |
|