Search
Search

LOC4324821| Gene Symbol: LOC4324821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763317.2 | XP_015618803.1 | 1 | 1184 | 117 | |
| GenBank mRNA | AK069147.1 | - | 1 | 1146 | 117 | |
| AK105929.1 | - | 1 | 1110 | 117 | ||
| AK106144.1 | - | 1 | 1140 | 117 | ||
| AK106186.1 | - | 1 | 1136 | 117 | ||
| AK289189.1 | - | 3038 | 117 | |||
Set Membership: |
|