Search
Search

LOC4331696| Gene Symbol: LOC4331696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776108.2 | XP_015631594.1 | 8 - 9 | 2708 | 145 | |
| GenBank mRNA | AK111595.1 | - | 8 - 9 | 2682 | 145 | |
| AY167575.1 | - | 8 - 9 | 2540 | 145 | ||
Set Membership: |
|