Search
Search

LOC4333004| Gene Symbol: LOC4333004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775863.2 | XP_015631349.1 | 3 - 4 | 382 | 76 | |
| XM_015775864.2 | XP_015631350.1 | 2 - 3 | 352 | 76 | ||
| XM_015775867.2 | XP_015631353.1 | 3 - 4 | 382 | 76 | ||
| XM_015775868.2 | XP_015631354.1 | 2 - 3 | 352 | 76 | ||
| XM_015775869.2 | XP_015631355.1 | 3 - 4 | 382 | 76 | ||
| XM_015775870.2 | XP_015631356.1 | 3 - 4 | 382 | 76 | ||
| XM_015775871.2 | XP_015631357.1 | 3 - 4 | 382 | 76 | ||
| XM_015775872.2 | XP_015631358.1 | 3 - 4 | 382 | 76 | ||
| XM_015775873.2 | XP_015631359.1 | 3 - 4 | 382 | 76 | ||
| GenBank mRNA | AK099666.1 | - | 2 - 3 | 220 | 76 | |
| AK100539.1 | - | 3 - 4 | 376 | 76 | ||
| AK105057.1 | - | 2 - 3 | 300 | 76 | ||
Set Membership: |
|