Search
Search

LOC4335926| Gene Symbol: LOC4335926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781502.2 | XP_015636988.1 | 10 - 11 | 1161 | 73 | |
| GenBank mRNA | AK062578.1 | - | 10 - 11 | 1062 | 73 | |
| AK065119.1 | - | 10 - 11 | 1116 | 73 | ||
| AK098870.1 | - | 10 - 11 | 1026 | 73 | ||
Set Membership: |
|