Search
Search

LOC4336212| Gene Symbol: LOC4336212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781218.2 | XP_015636704.1 | 1 - 2 | 1784 | 119 | |
| GenBank mRNA | AK100299.1 | - | 1 - 2 | 1780 | 119 | |
| AK102446.1 | - | 1 - 2 | 1568 | 119 | ||
Set Membership: |
|