Search
Search

LOC4339710| Gene Symbol: LOC4339710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784069.2 | XP_015639555.1 | 14 - 15 | 1361 | 151 | |
| GenBank mRNA | AK068594.1 | - | 14 - 15 | 1333 | 151 | |
Set Membership: |
|