Search
Search

LOC4339678| Gene Symbol: LOC4339678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785183.2 | XP_015640669.1 | 1 - 2 | 1431 | 50 | |
| GenBank mRNA | AK242382.1 | - | 1 - 2 | 86 | 50 | |
Set Membership: |
|