Search
Search

LOC4326133| Gene Symbol: LOC4326133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758013.2 | XP_015613499.1 | 15 - 16 | 3086 | 111 | |
| GenBank mRNA | AK121497.1 | - | 14 - 15 | 2490 | 111 | |
Set Membership: |
|