Search
Search

LOC4332849| Gene Symbol: LOC4332849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777516.2 | XP_015633002.1 | 1 - 2 | 157 | 80 | |
| GenBank mRNA | AK070491.1 | - | 1 - 2 | 139 | 80 | |
Set Membership: |
|