Search
Search

LOC4332788| Gene Symbol: LOC4332788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776691.2 | XP_015632177.1 | 13 - 14 | 2317 | 110 | |
| GenBank mRNA | AK060797.1 | - | 3 - 4 | 664 | 110 | |
| AK099406.1 | - | 13 - 14 | 2306 | 110 | ||
| AK102158.1 | - | 13 - 14 | 2307 | 110 | ||
| HQ895722.1 | - | 13 - 14 | 2255 | 110 | ||
| JN944363.1 | - | 13 - 14 | 2273 | 110 | ||
Set Membership: |
|