Search
Search

LOC4340517| Gene Symbol: LOC4340517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788490.2 | XP_015643976.1 | 3 - 4 | 867 | 75 | |
| GenBank mRNA | AK058664.1 | - | 3 - 4 | 869 | 75 | |
Set Membership: |
|