Search
Search

LOC4329567| Gene Symbol: LOC4329567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767950.2 | XP_015623436.1 | 2 - 3 | 312 | 99 | |
| GenBank mRNA | AK111881.1 | - | 2 - 3 | 316 | 99 | |
Set Membership: |
|