Search
Search

LOC4337821| Gene Symbol: LOC4337821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784917.2 | XP_015640403.1 | 9 - 10 | 1055 | 116 | |
| GenBank mRNA | AK058366.1 | - | 5 - 6 | 503 | 116 | |
| AK070081.1 | - | 9 - 10 | 1038 | 116 | ||
Set Membership: |
|