Search
Search

LOC4343970| Gene Symbol: LOC4343970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792079.2 | XP_015647565.1 | 1 - 2 | 1159 | 67 | |
| GenBank mRNA | AK103267.1 | - | 1 - 2 | 1166 | 67 | |
Set Membership: |
|