Search
Search

LOC9268640| Gene Symbol: LOC9268640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793441.2 | XP_015648927.1 | 1 - 2 | 571 | 96 | |
| GenBank mRNA | AK064847.1 | - | 1 - 2 | 573 | 96 | |
Set Membership: |
|