Search
Search

LOC4350787| Gene Symbol: LOC4350787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759640.2 | XP_015615126.1 | 1 - 2 | 412 | 119 | |
| GenBank mRNA | AK068669.1 | - | 1 - 2 | 382 | 119 | |
Set Membership: |
|