Search
Search

LOC4327302| Gene Symbol: LOC4327302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778150.2 | XP_015633636.1 | 7 - 8 | 771 | 78 | |
| GenBank mRNA | AB196736.1 | - | 10 - 11 | 1121 | 78 | |
| AK061360.1 | - | 7 - 8 | 740 | 78 | ||
| AK102360.1 | - | 7 - 8 | 778 | 78 | ||
Set Membership: |
|