Search
Search

LOC4337665| Gene Symbol: LOC4337665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784681.2 | XP_015640167.1 | 2 - 3 | 1583 | 112 | |
| GenBank mRNA | AK120892.1 | - | 2 - 3 | 1275 | 112 | |
| KJ206898.1 | - | 2 - 3 | 974 | 112 | ||
Set Membership: |
|