Search
Search

LOC4339847| Gene Symbol: LOC4339847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787542.2 | XP_015643028.1 | 21 - 22 | 2019 | 89 | |
| XM_015787543.2 | XP_015643029.1 | 20 - 21 | 1959 | 89 | ||
| XM_015787544.2 | XP_015643030.1 | 20 - 21 | 1956 | 89 | ||
| GenBank mRNA | AK072800.1 | - | 21 - 22 | 2014 | 89 | |
Set Membership: |
|