Search
Search

LOC4345771| Gene Symbol: LOC4345771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795235.2 | XP_015650721.1 | 2 - 3 | 1123 | 148 | |
| XM_015795236.2 | XP_015650722.1 | 2 - 3 | 1123 | 148 | ||
| GenBank mRNA | AK066516.1 | - | 2 - 3 | 1129 | 148 | |
| AK067364.1 | - | 4 - 5 | 752 | 148 | ||
| AK098966.1 | - | 2 - 3 | 1129 | 148 | ||
Set Membership: |
|