Search
Search

LOC4342350| Gene Symbol: LOC4342350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791506.2 | XP_015646992.1 | 6 - 7 | 850 | 74 | |
| GenBank mRNA | AK062102.1 | - | 1 - 2 | 69 | 74 | |
| AK065901.1 | - | 6 - 7 | 828 | 74 | ||
| AK072048.1 | - | 6 - 7 | 831 | 74 | ||
| AK099460.1 | - | 6 - 7 | 828 | 74 | ||
Set Membership: |
|