Search
Search

LOC4338718| Gene Symbol: LOC4338718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784946.2 | XP_015640432.1 | 1 - 2 | 638 | 121 | |
| GenBank mRNA | AK108949.1 | - | 1 - 2 | 292 | 121 | |
| X56787.1 | - | 1 - 2 | 627 | 121 | ||
Set Membership: |
|