Search
Search

OSBPL1A| Gene Symbol: OSBPL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121690.2 | XP_014977176.1 | 1197 | 93 | ||
| XM_015121691.2 | XP_014977177.1 | 1562 | 93 | |||
| XM_015121692.2 | XP_014977178.1 | 1088 | 93 | |||
| XM_015121693.2 | XP_014977179.1 | 1116 | 93 | |||
| XM_028837998.1 | XP_028693831.1 | 2419 | 93 | |||
| XM_028837999.1 | XP_028693832.1 | 2242 | 93 | |||
| XM_028838000.1 | XP_028693833.1 | 2236 | 93 | |||
| XM_028838001.1 | XP_028693834.1 | 2109 | 93 | |||
| XM_028838002.1 | XP_028693835.1 | 1951 | 93 | |||
| XM_028838003.1 | XP_028693836.1 | 1049 | 93 | |||
| GenBank mRNA | JU319707.1 | - | 1234 | 93 | ||
| JU319708.1 | - | 1229 | 93 | |||
| JU332805.1 | - | 2360 | 93 | |||
| JV706300.1 | - | 2354 | 93 | |||
Set Membership: |
|