Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh00402222_m1
See other ABLIM2 GE Assays ›
Gene Symbol:
ABLIM2
Gene Name:
actin binding LIM protein family member 2
Gene Aliases:
-
Chromosome Location:
Chr.5: 4777225 - 4974251 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh00402222_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ABLIM2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_002803994.3
XP_002804040.1
1020
67
XM_002803995.3
XP_002804041.1
1020
67
XM_002803996.3
XP_002804042.1
1020
67
XM_002803997.3
XP_002804043.1
1020
67
XM_002803998.3
XP_002804044.1
1019
67
XM_002803999.3
XP_002804045.1
1019
67
XM_015137963.2
XP_014993449.1
1020
67
XM_015137967.2
XP_014993453.1
1020
67
XM_015137984.2
XP_014993470.1
1021
67
XM_028848295.1
XP_028704128.1
919
67
XM_028848296.1
XP_028704129.1
919
67
XM_028848297.1
XP_028704130.1
1047
67
XM_028848298.1
XP_028704131.1
919
67
XM_028848299.1
XP_028704132.1
919
67
XM_028848300.1
XP_028704133.1
919
67
XM_028848301.1
XP_028704134.1
919
67
XM_028848302.1
XP_028704135.1
919
67
XM_028848303.1
XP_028704136.1
1041
67
XM_028848304.1
XP_028704137.1
1041
67
XM_028848305.1
XP_028704138.1
919
67
XM_028848306.1
XP_028704139.1
919
67
XM_028848307.1
XP_028704140.1
919
67
XM_028848308.1
XP_028704141.1
919
67
XM_028848309.1
XP_028704142.1
919
67
XM_028848310.1
XP_028704143.1
1044
67
XM_028848311.1
XP_028704144.1
919
67
XM_028848312.1
XP_028704145.1
919
67
XM_028848313.1
XP_028704146.1
1037
67
XM_028848314.1
XP_028704147.1
919
67
XM_028848315.1
XP_028704148.1
919
67
XM_028848316.1
XP_028704149.1
1044
67
XM_028848317.1
XP_028704150.1
1044
67
XM_028848318.1
XP_028704151.1
919
67
XM_028848319.1
XP_028704152.1
1037
67
XM_028848320.1
XP_028704153.1
1036
67
XM_028848321.1
XP_028704154.1
1037
67
XM_028848322.1
XP_028704155.1
1053
67
GenBank mRNA
JU336735.1
-
873
67
JU336736.1
-
873
67
JU477505.1
-
932
67
JU477506.1
-
932
67
JV048454.1
-
881
67
JV048455.1
-
881
67
JV631008.1
-
966
67
JV635590.1
-
866
67
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Macaca mulatta
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline