Search
Search

SCAI| Gene Symbol: SCAI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116845.2 | XP_014972331.2 | 447 | 100 | ||
| XM_015116846.2 | XP_014972332.1 | 441 | 100 | |||
| XM_028834849.1 | XP_028690682.1 | 477 | 100 | |||
| XM_028834850.1 | XP_028690683.1 | 441 | 100 | |||
| GenBank mRNA | JU331447.1 | - | 222 | 100 | ||
| JU485007.1 | - | 326 | 100 | |||
Set Membership: |
|