Search
Search

CSNK2A1| Gene Symbol: CSNK2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260877.1 | NP_001247806.1 | 416 | 141 | ||
| XM_015149293.2 | XP_015004779.1 | 560 | 141 | |||
| XM_015149294.2 | XP_015004780.1 | 350 | 141 | |||
| XM_015149295.2 | XP_015004781.1 | 279 | 141 | |||
| GenBank mRNA | JU333058.1 | - | 535 | 141 | ||
| JU333059.1 | - | 418 | 141 | |||
| JU474397.1 | - | 446 | 141 | |||
| JV046251.1 | - | 416 | 141 | |||
| JV606622.1 | - | 440 | 141 | |||
Set Membership: |
|