Search
Search

HMGCLL1| Gene Symbol: HMGCLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105003.4 | XP_001105003.3 | 743 | 72 | ||
| XM_002803774.2 | XP_002803820.2 | 693 | 72 | |||
| XM_028847467.1 | XP_028703300.1 | 507 | 72 | |||
| XM_028847468.1 | XP_028703301.1 | 533 | 72 | |||
| XM_028847469.1 | XP_028703302.1 | 674 | 72 | |||
| GenBank mRNA | JV717284.1 | - | 469 | 72 | ||
Set Membership: |
|