Search
Search

ATRNL1| Gene Symbol: ATRNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808513.3 | XP_002808559.2 | 1475 | 97 | ||
| XM_015148255.2 | XP_015003741.1 | 1467 | 97 | |||
| XM_015148256.2 | XP_015003742.1 | 1467 | 97 | |||
| XM_015148257.2 | XP_015003743.1 | 1467 | 97 | |||
| XM_015148258.2 | XP_015003744.1 | 816 | 97 | |||
| GenBank mRNA | JU333906.1 | - | 1367 | 97 | ||
| JV612575.1 | - | 1468 | 97 | |||