Search
Search

CRB2| Gene Symbol: CRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083811.4 | XP_001083811.3 | 503 | 132 | ||
| XM_015116889.2 | XP_014972375.2 | 503 | 132 | |||
| GenBank mRNA | JU325310.1 | - | 364 | 132 | ||
| JV614123.1 | - | 354 | 132 | |||
Set Membership: |
|