Search
Search

NRDC| Gene Symbol: NRDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139671.2 | XP_014995157.1 | 1254 | 96 | ||
| XM_015139676.2 | XP_014995162.1 | 1570 | 96 | |||
| XM_028833839.1 | XP_028689672.1 | 718 | 96 | |||
| GenBank mRNA | JU334699.1 | - | 1068 | 96 | ||
| JU475746.1 | - | 1073 | 96 | |||
| JV047179.1 | - | 1088 | 96 | |||
| JV565132.1 | - | 1302 | 96 | |||
| JV652576.1 | - | 1099 | 96 | |||
Set Membership: |
|