Search
Search

FUBP3| Gene Symbol: FUBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257743.1 | NP_001244672.1 | 1134 | 68 | ||
| XM_015116533.2 | XP_014972019.1 | 1217 | 68 | |||
| XM_015116534.2 | XP_014972020.1 | 1220 | 68 | |||
| XM_015116536.2 | XP_014972022.1 | 4253 | 68 | |||
| XM_015116537.2 | XP_014972023.1 | 4311 | 68 | |||
| GenBank mRNA | JU321970.1 | - | 1134 | 68 | ||
| JU473485.1 | - | 1120 | 68 | |||
| JV048613.1 | - | 1153 | 68 | |||
Set Membership: |
|