Search
Search

IQGAP1| Gene Symbol: IQGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143023.2 | XP_014998509.1 | 1034 | 82 | ||
| GenBank mRNA | JU336926.1 | - | 930 | 82 | ||
| JU477661.1 | - | 872 | 82 | |||
| JV048590.1 | - | 857 | 82 | |||
| JV597235.1 | - | 920 | 82 | |||
Set Membership: |
|