Search
Search

ASXL1| Gene Symbol: ASXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149252.2 | XP_015004738.2 | 590 | 70 | ||
| XM_015149253.2 | XP_015004739.2 | 1459 | 70 | |||
| XM_015149254.2 | XP_015004740.2 | 1456 | 70 | |||
| XM_015149258.2 | XP_015004744.2 | 684 | 70 | |||
| XM_015149259.2 | XP_015004745.2 | 1572 | 70 | |||
| XM_028828302.1 | XP_028684135.1 | 438 | 70 | |||
| GenBank mRNA | JU323366.1 | - | 518 | 70 | ||
| JU474538.1 | - | 498 | 70 | |||
| JV044726.1 | - | 494 | 70 | |||
| JV573326.1 | - | 551 | 70 | |||
| JV637395.1 | - | 501 | 70 | |||
Set Membership: |
|