Search
Search

DNAJC6| Gene Symbol: DNAJC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089936.3 | XP_001089936.1 | 2268 | 55 | ||
| XM_001090057.4 | XP_001090057.1 | 2324 | 55 | |||
| XM_015141751.2 | XP_014997237.1 | 2116 | 55 | |||
| XM_015141756.2 | XP_014997242.1 | 2369 | 55 | |||
| XM_015141774.2 | XP_014997260.1 | 2217 | 55 | |||
| XM_015141779.2 | XP_014997265.1 | 1990 | 55 | |||
| XM_028832824.1 | XP_028688657.1 | 2251 | 55 | |||
| GenBank mRNA | JU337344.1 | - | 2105 | 55 | ||
| JV564443.1 | - | 2107 | 55 | |||
| JV634922.1 | - | 2102 | 55 | |||
Set Membership: |
|