Search
Search

COG7| Gene Symbol: COG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125796.2 | XP_014981282.1 | 1695 | 92 | ||
| GenBank mRNA | JU477809.1 | - | 1652 | 92 | ||
| JU477810.1 | - | 1652 | 92 | |||
| JV636449.1 | - | 1562 | 92 | |||
| JV726813.1 | - | 1641 | 92 | |||