Search
Search

ZNF76| Gene Symbol: ZNF76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136192.2 | XP_014991678.1 | 923 | 80 | ||
| XM_015136193.2 | XP_014991679.1 | 930 | 80 | |||
| XM_015136194.2 | XP_014991680.1 | 840 | 80 | |||
| XM_015136195.2 | XP_014991681.1 | 912 | 80 | |||
| XM_015136196.2 | XP_014991682.1 | 923 | 80 | |||
| XM_015136197.2 | XP_014991683.1 | 923 | 80 | |||
| XM_015136198.2 | XP_014991684.1 | 840 | 80 | |||
| GenBank mRNA | JU322480.1 | - | 783 | 80 | ||
| JU472056.1 | - | 817 | 80 | |||
| JV048196.1 | - | 819 | 80 | |||
| JV592945.1 | - | 797 | 80 | |||
Set Membership: |
|