Search
Search

HFE| Gene Symbol: HFE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260576.2 | NP_001247505.1 | 84 | 106 | ||
| XM_015135625.2 | XP_014991111.2 | 229 | 106 | |||
| GenBank mRNA | JU334244.1 | - | 84 | 106 | ||
| JU334245.1 | - | 84 | 106 | |||
Set Membership: |
|