Search
Search

CERS5| Gene Symbol: CERS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001102810.4 | XP_001102810.3 | 834 | 86 | ||
| XM_015151690.2 | XP_015007176.2 | 774 | 86 | |||
| XM_015151691.2 | XP_015007177.2 | 774 | 86 | |||
| XM_015151692.2 | XP_015007178.1 | 774 | 86 | |||
| XM_028829467.1 | XP_028685300.1 | 774 | 86 | |||
| XM_028829468.1 | XP_028685301.1 | 774 | 86 | |||
| GenBank mRNA | JU337024.1 | - | 561 | 86 | ||
| JV075212.1 | - | 494 | 86 | |||
Set Membership: |
|