Search
Search

G2E3| Gene Symbol: G2E3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194652.2 | NP_001181581.2 | 891 | 67 | ||
| XM_001114675.4 | XP_001114675.2 | 1264 | 67 | |||
| XM_015143477.2 | XP_014998963.2 | 899 | 67 | |||
| XM_015143478.2 | XP_014998964.2 | 960 | 67 | |||
| XM_028850545.1 | XP_028706378.1 | 773 | 67 | |||
| XM_028850546.1 | XP_028706379.1 | 857 | 67 | |||
| GenBank mRNA | JU472603.1 | - | 856 | 67 | ||
| JU472604.1 | - | 856 | 67 | |||
| JU472605.1 | - | 856 | 67 | |||
Set Membership: |
|