Search
Search

EGR1| Gene Symbol: EGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266878.1 | NP_001253807.1 | 519 | 71 | ||
| GenBank mRNA | JU327816.1 | - | 60 | 71 | ||
| JU327817.1 | - | 60 | 71 | |||
| JU474588.1 | - | 512 | 71 | |||
| JV046371.1 | - | 519 | 71 | |||
| JV527786.1 | - | 362 | 71 | |||
Set Membership: |
|