Search
Search

LUM| Gene Symbol: LUM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266327.1 | NP_001253256.1 | 98 | 111 | ||
| GenBank mRNA | JU324449.1 | - | 98 | 111 | ||
| JU473601.1 | - | 94 | 111 | |||
| JV044750.1 | - | 98 | 111 | |||
Set Membership: |
|