Search
Search

GRM3| Gene Symbol: GRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107588.4 | XP_001107588.1 | 3547 | 82 | ||
| XM_015134166.2 | XP_014989652.1 | 3136 | 82 | |||
| XM_028846121.1 | XP_028701954.1 | 2680 | 82 | |||
| XM_028846122.1 | XP_028701955.1 | 2939 | 82 | |||
| XM_028846123.1 | XP_028701956.1 | 2083 | 82 | |||
| GenBank mRNA | JV567927.1 | - | 3114 | 82 | ||
Set Membership: |
|